2-Amino-6-fluorpyridin

Lieferant: COMBI-BLOCKS
Achtung

OS-7692-5G OS-7692-1G OS-7692-25G
COBBOS-7692-5GEA 75.8 EUR
COBBOS-7692-5G COBBOS-7692-1G COBBOS-7692-25G
2-Amino-6-fluorpyridin
2-Amino-6-fluorpyridin
Formel: C₅H₅FN₂
Molecular Weight: 112,11 g/mol
MDL: MFCD04114200
CAS-Nummer: 1597-32-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR