2-Amino-6-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methyl-3-nitropyridin-2-amin, 2-Amino-3-nitro-6-picolin, 6-Methyl-3-nitro-2-pyridylamin

OT-0182-1G OT-0182-25G OT-0182-5G
COBBOT-0182-1GEA 26 EUR
COBBOT-0182-1G COBBOT-0182-25G COBBOT-0182-5G
2-Amino-6-methyl-3-nitropyridin
2-Amino-6-methyl-3-nitropyridin
Formel: C₆H₇N₃O₂
Molecular Weight: 153.14 g/mol
MDL: MFCD00047443
CAS-Nummer: 21901-29-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR