6-Methylanthranilsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Amino-6-methylbenzoic acid
CA-4023-1G
CA-4023-5G
CA-4023-25G
COBBCA-4023-1GEA
25.5
EUR
COBBCA-4023-1G
COBBCA-4023-5G
COBBCA-4023-25G
6-Methylanthranilsäure
6-Methylanthranilsäure
Formel:
C₈H₉NO₂ Molecular Weight: 151.16 g/mol Siedepunkt: 315 Schmelzpunkt: 125…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007809 CAS-Nummer: 4389-50-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...