2-Aminobenzimidazol 99+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzimidazol-2-ylamin, 2-Amino-1H-benzimidazol
174020250
174021000
ACRO174020250EA
85.2
EUR
ACRO174020250
ACRO174021000
2-Aminobenzimidazol 99+%
2-Aminobenzimidazole
Formel:
C₇H₇N₃ Molecular Weight: 133,15 g/mol Siedepunkt: 381 °C (1013 hPa) Schmelzpunkt: 228…232 °C |
MDL:
MFCD00005596 CAS-Nummer: 934-32-7 EINECS: 213-280-6 |
Spezifikation Testergebnisse
Appearance | Yellow to beige to light brown Crystalline powder or crystals or flakes (May darken on storage) |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥99.0 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...