2-Aminobenzolsulfonamid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Anilin-2-sulfonsäureamid, 2-Aminobenzolsulfonsäureamid
M-Clarity™ Qualitätsstufe = MQ100
245887-25G
245887-5G
SIAL245887-25GEA
188
EUR
SIAL245887-25G
SIAL245887-5G
2-Aminobenzolsulfonamid, Sigma-Aldrich®
2-Aminobenzolsulfonamid
Formel:
C₆H₈N₂O₂S Molecular Weight: 172,21 g/mol |
MDL:
MFCD00007932 CAS-Nummer: 3306-62-5 EINECS: 221-988-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...