2-Aminophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Aminobenzolboronsäure, Dihydroxy(2-aminophenyl)boran, (2-Aminophenyl)metaborsäure

BB-2139-1G BB-2139-5G
COBBBB-2139-1GEA 0 EUR
COBBBB-2139-1G COBBBB-2139-5G
2-Aminophenylboronsäure
2-Aminophenylboronsäure
Formel: C₆H₈BNO₂
Molecular Weight: 136.95 g/mol
MDL: MFCD01074645
CAS-Nummer: 5570-18-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR