2-Aminopyrazin

Lieferant: COMBI-BLOCKS

Synonyme: Aminopyrazin, Pyrazin-2-amin

OS-7492-25G OS-7492-100G
COBBOS-7492-25GEA 38.2 EUR
COBBOS-7492-25G COBBOS-7492-100G
2-Aminopyrazin
2-Aminopyrazine
Formel: C₄H₅N₃
Molecular Weight: 95,1 g/mol
Siedepunkt: 203 °C (1013 hPa)
Schmelzpunkt: 117…120 °C
MDL: MFCD00006137
CAS-Nummer: 5049-61-6
EINECS: 225-748-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR