2-Adamantylamin Hydrochlorid ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: 2-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, 2-Aminoadamantan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-2-ylaminhydrochlorid, 2-Adamantanammoniumchlorid
A17140.03 A17140.06 A17140.14
A17140.03EA 24.4 EUR
A17140.03 A17140.06 A17140.14
2-Adamantylamin Hydrochlorid ≥98%
2-Adamantylamin Hydrochlorid
Purity: 98+% CAS No.: 10523-68-9 Molecular Formula: C10H17N.HCl Formula Weight: 187.71 Melting Point: 300° Beilstein Reference: 4297901
Formel: C₁₀H₁₈ClN
Molecular Weight: 187,71 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074743
CAS-Nummer: 10523-68-9
EINECS: 234-074-2