2-Adamantylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, 2-Aminoadamantan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-2-ylaminhydrochlorid, 2-Adamantanammoniumchlorid

M-Clarity™ Qualitätsstufe = MQ100

153818-25G 153818-5G
SIAL153818-25GEA 265 EUR
SIAL153818-25G SIAL153818-5G
2-Adamantylamin Hydrochlorid, Sigma-Aldrich®
2-Adamantylamin Hydrochlorid
Formel: C₁₀H₁₈ClN
Molecular Weight: 187,71 g/mol
Schmelzpunkt: 300 °C
MDL: MFCD00074743
CAS-Nummer: 10523-68-9
EINECS: 234-074-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR