2-Adamantylamin Hydrochlorid

Lieferant: CHEMPUR

Synonyme: 2-Aminotricyclo[3.3.1.1(3,7)]decan Hydrochlorid, 2-Aminoadamantan Hydrochlorid, Tricyclo[3.3.1.1(3,7)]dec-2-ylaminhydrochlorid, 2-Adamantanammoniumchlorid

BTB12185.250 BTB12185.1
CHMPBTB12185.250EA 0 EUR
CHMPBTB12185.250 MAYBBTB12185.1
2-Adamantylamin Hydrochlorid
2-Adamantylamin Hydrochlorid
Formel: C₁₀H₁₈ClN
Molecular Weight: 187,71 g/mol
Schmelzpunkt: 300 °C
MDL: MFCD00074743
CAS-Nummer: 10523-68-9
EINECS: 234-074-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR