2-Brom-1-methylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-1-methyl-1H-imidazol

HI-1040-5G HI-1040-1G
COBBHI-1040-5GEA 470 EUR
COBBHI-1040-5G COBBHI-1040-1G
2-Brom-1-methylimidazol
2-Bromo-1-methylimidazole
Formel: C₄H₅BrN₂
Molecular Weight: 161 g/mol
MDL: MFCD02179525
CAS-Nummer: 16681-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR