2-Brom-1-methylimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-1-methyl-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

639850-1G 639850-5G
SIAL639850-1GEA 79 EUR
SIAL639850-1G SIAL639850-5G
2-Brom-1-methylimidazol, Sigma-Aldrich®
2-Bromo-1-methylimidazole
Formel: C₄H₅BrN₂
Molecular Weight: 161 g/mol
MDL: MFCD02179525
CAS-Nummer: 16681-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR