2-Brom-3,5-dichlorpyridin

Lieferant: COMBI-BLOCKS

CA-4307-5G CA-4307-25G CA-4307-1G CA-4307-100G
COBBCA-4307-5GEA 0 EUR
COBBCA-4307-5G COBBCA-4307-25G COBBCA-4307-1G COBBCA-4307-100G
2-Brom-3,5-dichlorpyridin
2-Bromo-3,5-dichloropyridine
Formel: C₅H₂BrCl₂N
MDL: MFCD00233990
CAS-Nummer: 14482-51-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR