2-Brom-3-hydroxy-4-methoxybenzaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

478075-25G 478075-5G
SIAL478075-25GEA 317 EUR
SIAL478075-25G SIAL478075-5G
2-Brom-3-hydroxy-4-methoxybenzaldehyd, Sigma-Aldrich®
2-Bromo-3-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇BrO₃
Molecular Weight: 231,05 g/mol
MDL: MFCD02380230
CAS-Nummer: 2973-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR