2-Brom-3-methylpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Bromo-3-picoline
OS-7437-5G
OS-7437-25G
OS-7437-100G
COBBOS-7437-5GEA
26.2
EUR
COBBOS-7437-5G
COBBOS-7437-25G
COBBOS-7437-100G
2-Brom-3-methylpyridin
2-Brom-3-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol |
MDL:
MFCD00239380 CAS-Nummer: 3430-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...