2-Brom-3-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brompyridin-3-boronsäure, 2-Brom-3-pyridinylboronsäure

BB-2705-5G BB-2705-1G
COBBBB-2705-5GEA 208 EUR
COBBBB-2705-5G COBBBB-2705-1G
2-Brom-3-pyridinboronsäure
2-Bromo-3-pyridineboronic acid
Formel: C₅H₅BBrNO₂
Molecular Weight: 201,81 g/mol
MDL: MFCD04113805
CAS-Nummer: 452972-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR