2-Bromnicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-3-pyridincarbonsäure, 2-Brompyridin-3-carbonsäure

OS-7435-5G OS-7435-25G OS-7435-100G
COBBOS-7435-5GEA 26.2 EUR
COBBOS-7435-5G COBBOS-7435-25G COBBOS-7435-100G
2-Bromnicotinsäure
2-Bromnicotinsäure
Formel: C₆H₄BrNO₂
Molecular Weight: 202.01 g/mol
MDL: MFCD01646068
CAS-Nummer: 35905-85-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR