2-Brom-4,5-dichlorimidazol
Lieferant: Thermo Scientific
Synonyme:
2-Brom-4,5-dichlor-1H-imidazol
KM01467DA
KM01467EA
MAYBKM01467DAEA
0
EUR
MAYBKM01467DA
MAYBKM01467EA
2-Brom-4,5-dichlorimidazol
2-Bromo-4,5-dichloroimidazole
Formel:
C₃HBrCl₂N₂ Molecular Weight: 215.86 g/mol |
MDL:
MFCD00067894 CAS-Nummer: 16076-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...