2-Brom-4,5-dichlorimidazol
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-4,5-dichlor-1H-imidazol
CFF404.10
CFF404.50
017427-10G
017427-25G
017427-50G
CHMPCFF404.10EA
532
EUR
CHMPCFF404.10
CHMPCFF404.50
FLUO017427-10G
FLUO017427-25G
FLUO017427-50G
2-Brom-4,5-dichlorimidazol
2-Bromo-4,5-dichloroimidazole
Formel:
C₃HBrCl₂N₂ Molecular Weight: 215.86 g/mol |
MDL:
MFCD00067894 CAS-Nummer: 16076-27-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...