2-Brom-4,5-dichlorimidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Brom-4,5-dichlor-1H-imidazol
M-Clarity™ Qualitätsstufe = MQ100
548723-1G
548723-5G
SIAL548723-1GEA
63.6
EUR
SIAL548723-1G
SIAL548723-5G
2-Brom-4,5-dichlorimidazol, Sigma-Aldrich®
2-Bromo-4,5-dichloroimidazole
Formel:
C₃HBrCl₂N₂ Molecular Weight: 215,86 g/mol |
MDL:
MFCD00067894 CAS-Nummer: 16076-27-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...