6-Bromveratraldehyd

Lieferant: COMBI-BLOCKS

Synonyme: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde

OS-7684-5G OS-7684-25G
COBBOS-7684-5GEA 25.9 EUR
COBBOS-7684-5G COBBOS-7684-25G
6-Bromveratraldehyd
6-Bromveratraldehyd
Formel: C₉H₉BrO₃
Molecular Weight: 245.07 g/mol
Siedepunkt: 300
Schmelzpunkt: 151
MDL: MFCD00003301
CAS-Nummer: 5392-10-9
EINECS: 226-390-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR