2-Brom-4,5-dimethoxybenzoesäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 6-Bromveratrumsäure, 6-Bromveratrinsäure

CA-4967-5G CA-4967-25G
COBBCA-4967-5GEA 50.9 EUR
COBBCA-4967-5G COBBCA-4967-25G
2-Brom-4,5-dimethoxybenzoesäure
2-Brom-4,5-dimethoxybenzoesäure
Formel: C₉H₉BrO₄
Molecular Weight: 261,07 g/mol
MDL: MFCD00017542
CAS-Nummer: 6286-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR