2-Brom-4-chlorbenzoesäure

Lieferant: COMBI-BLOCKS

CA-4589-5G CA-4589-25G
COBBCA-4589-5GEA 25.9 EUR
COBBCA-4589-5G COBBCA-4589-25G
2-Brom-4-chlorbenzoesäure
2-Bromo-4-chlorobenzoic acid
Formel: C₇H₄BrClO₂
Molecular Weight: 235,46 g/mol
MDL: MFCD00672930
CAS-Nummer: 936-08-3
UN: 2928
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR