2-Brom-4-fluorbenzaldehyd
Lieferant: COMBI-BLOCKS
OS-7685-25G
OS-7685-100G
COBBOS-7685-25GEA
0
EUR
COBBOS-7685-25G
COBBOS-7685-100G
2-Brom-4-fluorbenzaldehyd
2-Bromo-4-fluorobenzaldehyde
Formel:
C₇H₄BrFO |
MDL:
MFCD00672923 CAS-Nummer: 59142-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...