2-Brom-4-fluorbenzolsulfonylchlorid ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-Brom-4-fluorbenzolsulfonsäurechlorid, 2-Brom-4-fluorbenzolsulfochlorid
L19713.03
L19713.06
L19713.03EA
26.6
EUR
L19713.03
L19713.06
2-Brom-4-fluorbenzolsulfonylchlorid ≥97%
2-Brom-4-fluorbenzolsulfonylchlorid
Purity: 97%
CAS No.: 351003-45-7
Molecular Formula: C6H3BrClFO2S
Formula Weight: 273.51
Refractive Index: 1.5780
Boiling Point: 264-265°
UN No.: 2920
MDL No.: MFCD03094149
Formel:
C₆H₃BrClFO₂S Molecular Weight: 273,51 g/mol Siedepunkt: 264…265 °C Dichte: 1,870 Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094149 CAS-Nummer: 351003-45-7 UN: 2920 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...