2-Brom-4-methylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-4-picoline
PY-7764-100G
PY-7764-25G
PY-7764-5G
COBBPY-7764-100GEA
127
EUR
COBBPY-7764-100G
COBBPY-7764-25G
COBBPY-7764-5G
2-Brom-4-methylpyridin
2-Brom-4-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol Siedepunkt: 42…44 °C (0.6 mmHg) Schmelzpunkt: 218…228 °C Dichte: 1.545 g/cm³ (20 °C) |
MDL:
MFCD00082590 CAS-Nummer: 4926-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...