2-Brom-5-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Brom-5-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Brom-5-(trifluormethyl)benzolsulfochlorid, 2-Brom-5-(trifluormethyl)benzolsulfonsäurechlorid
M-Clarity™ Qualitätsstufe = MQ100
558435-1G
558435-5G
SIAL558435-1GEA
56.2
EUR
SIAL558435-1G
SIAL558435-5G
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid, Sigma-Aldrich®
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃BrClF₃O₂S Molecular Weight: 323,52 g/mol Siedepunkt: 230…231 °C (1013 hPa) |
MDL:
MFCD03094395 CAS-Nummer: 176225-08-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...