2-Brom-5-hydroxy-4-methoxybenzaldehyd

Lieferant: CHEMPUR

FL13447.POR 013447-5G
CHMPFL13447.POREA 0 EUR
CHMPFL13447.POR FLUO013447-5G
2-Brom-5-hydroxy-4-methoxybenzaldehyd
2-Bromo-5-hydroxy-4-methoxybenzaldehyde
Formel: C₈H₇BrO₃
Molecular Weight: 231.05 g/mol
MDL: MFCD00195552
CAS-Nummer: 2973-59-3
EINECS: 426-540-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR