2-Brom-5-nitropyridin

Lieferant: COMBI-BLOCKS
Achtung

OT-0312-5G OT-0312-25G OT-0312-100G
COBBOT-0312-5GEA 38.8 EUR
COBBOT-0312-5G COBBOT-0312-25G COBBOT-0312-100G
2-Brom-5-nitropyridin
2-Bromo-5-nitropyridine
Formel: C₅H₃BrN₂O₂
Molecular Weight: 203 g/mol
MDL: MFCD00006222
CAS-Nummer: 4487-59-6
EINECS: 224-777-2
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR