2-Brom-5-nitrothiazol

Lieferant: COMBI-BLOCKS

HI-1160-5G HI-1160-25G HI-1160-1G
COBBHI-1160-5GEA 125 EUR
COBBHI-1160-5G COBBHI-1160-25G COBBHI-1160-1G
2-Brom-5-nitrothiazol
2-Bromo-5-nitrothiazole
Formel: C₃HBrN₂O₂S
Molecular Weight: 209.02 g/mol
Schmelzpunkt: 93
Dichte: 2.295 g/cm³ (20 °C)
MDL: MFCD00005317
CAS-Nummer: 3034-48-8
EINECS: 221-226-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR