2-Brom-6-chlorpyridin

Lieferant: COMBI-BLOCKS

OS-7403-1G OS-7403-25G OS-7403-100G OS-7403-5G
COBBOS-7403-1GEA 25.5 EUR
COBBOS-7403-1G COBBOS-7403-25G COBBOS-7403-100G COBBOS-7403-5G
2-Brom-6-chlorpyridin
2-Bromo-6-chloropyridine
Formel: C₅H₃BrClN
Molecular Weight: 192.44 g/mol
MDL: MFCD00181262
CAS-Nummer: 5140-72-7
EINECS: 225-904-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR