2-Brom-6-fluorbenzaldehyd
Lieferant: COMBI-BLOCKS
OS-7650-5G
OS-7650-25G
OS-7650-1G
COBBOS-7650-5GEA
50.7
EUR
COBBOS-7650-5G
COBBOS-7650-25G
COBBOS-7650-1G
2-Brom-6-fluorbenzaldehyd
2-Bromo-6-fluorobenzaldehyde
Formel:
C₇H₄BrFO Molecular Weight: 203,01 g/mol |
MDL:
MFCD03407341 CAS-Nummer: 360575-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...