2-Brom-6-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-7708-1G OS-7708-25G OS-7708-5G
COBBOS-7708-1GEA 0 EUR
COBBOS-7708-1G COBBOS-7708-25G COBBOS-7708-5G
2-Brom-6-fluorpyridin
2-Bromo-6-fluoropyridine
Formel: C₅H₃BrFN
MDL: MFCD03095258
CAS-Nummer: 144100-07-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR