6-Brom-o-toluylsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: 2-Bromo-6-methylbenzoic acid

CA-4005-1G CA-4005-5G CA-4005-25G
COBBCA-4005-1GEA 51.3 EUR
COBBCA-4005-1G COBBCA-4005-5G COBBCA-4005-25G
6-Brom-o-toluylsäure
6-Brom-o-toluylsäure
Formel: C₈H₇BrO₂
Molecular Weight: 215,05 g/mol
MDL: MFCD01310788
CAS-Nummer: 90259-31-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR