2-Brom-6-methylpyridin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
6-Bromo-2-picoline, 2-Bromo-6-picoline
OS-7438-25G
OS-7438-100G
COBBOS-7438-25GEA
51.3
EUR
COBBOS-7438-25G
COBBOS-7438-100G
2-Brom-6-methylpyridin
2-Brom-6-methylpyridin
Formel:
C₆H₆BrN Molecular Weight: 172.02 g/mol |
MDL:
MFCD00040743 CAS-Nummer: 5315-25-3 EINECS: 226-173-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...