2-Brombenzamidin Hydrochlorid
Lieferant: CHEMPUR
Synonyme:
2-Bromphenylamidin Hydrochlorid, 2-Brombenzoesäureamidin Hydrochlorid
12-0054.500
12-0054.1
CHMP12-0054.500EA
0
EUR
CHMP12-0054.500
CHMP12-0054.1
2-Brombenzamidin Hydrochlorid
2-Brombenzamidin Hydrochlorid
Formel:
C₇H₈BrClN₂ Molecular Weight: 235.51 g/mol |
MDL:
MFCD04114438 CAS-Nummer: 57075-82-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...