2-Brombenzolsulfonamid

Lieferant: Thermo Scientific

Synonyme: 2-Brombenzolsulfonsäureamid

TL00539DA TL00539EA
MAYBTL00539DAEA 0 EUR
MAYBTL00539DA MAYBTL00539EA
2-Brombenzolsulfonamid
2-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00173662
CAS-Nummer: 92748-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR