2-Brombenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brombenzolsulfonsäureamid

HC-6577-5G HC-6577-25G HC-6577-1G
COBBHC-6577-5GEA 98.3 EUR
COBBHC-6577-5G COBBHC-6577-25G COBBHC-6577-1G
2-Brombenzolsulfonamid
2-Brombenzolsulfonamid
Formel: C₆H₆BrNO₂S
Molecular Weight: 236,09 g/mol
MDL: MFCD00173662
CAS-Nummer: 92748-09-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR