2-Brombenzolsulfonamid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Brombenzolsulfonsäureamid
M-Clarity™ Qualitätsstufe = MQ100
644609-1G
644609-5G
SIAL644609-1GEA
32.4
EUR
SIAL644609-1G
SIAL644609-5G
2-Brombenzolsulfonamid, Sigma-Aldrich®
2-Brombenzolsulfonamid
Formel:
C₆H₆BrNO₂S Molecular Weight: 236,09 g/mol |
MDL:
MFCD00173662 CAS-Nummer: 92748-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...