2-Bromchinolin

Lieferant: COMBI-BLOCKS
Achtung

PY-7953-5G PY-7953-1G PY-7953-100G
COBBPY-7953-5GEA 130 EUR
COBBPY-7953-5G COBBPY-7953-1G COBBPY-7953-100G
2-Bromchinolin
2-Bromoquinoline
Formel: C₉H₆BrN
Molecular Weight: 208,06 g/mol
MDL: MFCD00234480
CAS-Nummer: 2005-43-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR