2-Bromfuran
Lieferant: Thermo Scientific
CC02010.250
CC02010DA
CC02010.0001
MAYBCC02010.250EA
0
EUR
MAYBCC02010.250
MAYBCC02010DA
ABCRCC02010.0001
2-Bromfuran
2-Bromofuran
Formel:
C₄H₃BrO |
MDL:
MFCD00216609 CAS-Nummer: 584-12-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...