2-Bromimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brom-1H-imidazol

HI-1041-5G HI-1041-1G
COBBHI-1041-5GEA 264 EUR
COBBHI-1041-5G COBBHI-1041-1G
2-Bromimidazol
2-Bromoimidazole
Formel: C₃H₃BrN₂
Molecular Weight: 146,97 g/mol
MDL: MFCD02179526
CAS-Nummer: 16681-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR