2-Bromimidazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Brom-1H-imidazol

M-Clarity™ Qualitätsstufe = MQ100

666521-1G 666521-250MG
SIAL666521-1GEA 147 EUR
SIAL666521-1G SIAL666521-250MG
2-Bromimidazol, Sigma-Aldrich®
2-Bromoimidazole
Formel: C₃H₃BrN₂
Molecular Weight: 146,97 g/mol
MDL: MFCD02179526
CAS-Nummer: 16681-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR