2-Chlor-1-methylimidazol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Chlor-1-methyl-1H-imidazol

H51867.MD H51867.03
H51867.MDEA 72.6 EUR
H51867.MD H51867.03
2-Chlor-1-methylimidazol ≥97%
2-Chloro-1-methylimidazole

Purity: 97+% CAS No.: 253453-91-7 Molecular Formula: C4H5ClN2 Formula Weight: 116.55 Melting Point: -10° MDL No.: MFCD02179531

Formel: C₄H₅ClN₂
Molecular Weight: 116,55 g/mol
Schmelzpunkt: –10 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179531
CAS-Nummer: 253453-91-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR