2-Chlor-3-fluorpyridin

Lieferant: COMBI-BLOCKS

OS-7515-5G OS-7515-25G OS-7515-100G
COBBOS-7515-5GEA 26.2 EUR
COBBOS-7515-5G COBBOS-7515-25G COBBOS-7515-100G
2-Chlor-3-fluorpyridin
2-Chloro-3-fluoropyridine
Formel: C₅H₃ClFN
Molecular Weight: 131.54 g/mol
MDL: MFCD03095302
CAS-Nummer: 17282-04-1
UN: 2810
ADR: 6.1 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR