2-Chlor-3-methoxypyridin

Lieferant: COMBI-BLOCKS
Achtung

PY-1445-1G PY-1445-5G
COBBPY-1445-1GEA 38.2 EUR
COBBPY-1445-1G COBBPY-1445-5G
2-Chlor-3-methoxypyridin
2-Chloro-3-methoxypyridine
Formel: C₆H₆ClNO
Molecular Weight: 143,57 g/mol
MDL: MFCD03426022
CAS-Nummer: 52605-96-6
EINECS: 258-039-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR