2-Chlor-3-picolin
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2-Chlor-β-picolin, 2-Chlor-3-methylpyridin
OS-7516-5G
OS-7516-100G
OS-7516-25G
COBBOS-7516-5GEA
25.9
EUR
COBBOS-7516-5G
COBBOS-7516-100G
COBBOS-7516-25G
2-Chlor-3-picolin
2-Chlor-3-picolin
Formel:
C₆H₆ClN Molecular Weight: 127.57 g/mol |
MDL:
MFCD00234177 CAS-Nummer: 18368-76-8 EINECS: 242-242-1 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...