2-Chlor-3-methylthiophen
Lieferant: COMBI-BLOCKS
OS-7472-5G
OS-7472-25G
OS-7472-100G
COBBOS-7472-5GEA
0
EUR
COBBOS-7472-5G
COBBOS-7472-25G
COBBOS-7472-100G
2-Chlor-3-methylthiophen
2-Chloro-3-methylthiophene
Formel:
C₅H₅ClS Siedepunkt: 153…157 °C (1013 hPa) Dichte: 1,2281 g/cm³ (25 °C) |
MDL:
MFCD00130083 CAS-Nummer: 14345-97-2 EINECS: 238-296-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...