2-Chlor-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Chlor-3-pyridinylboronsäure, 2-Chlorpyridin-3-boronsäure
BB-2100-5G
BB-2100-25G
BB-2100-1G
COBBBB-2100-5GEA
50.8
EUR
COBBBB-2100-5G
COBBBB-2100-25G
COBBBB-2100-1G
2-Chlor-3-pyridinboronsäure
2-Chloro-3-pyridineboronic acid
Formel:
C₅H₅BClNO₂ Molecular Weight: 157.36 g/mol |
MDL:
MFCD03094997 CAS-Nummer: 381248-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...