2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-4-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Chlor-4-(trifluormethyl)benzolsulfochlorid, 2-Chlor-4-(trifluormethyl)benzolsulfonsäurechlorid
004536-1G
004536-25G
004536-5G
FLUO004536-1GEA
45.9
EUR
FLUO004536-1G
FLUO004536-25G
FLUO004536-5G
2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
2-Chlor-4-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃Cl₂F₃O₂S Siedepunkt: 70…71 °C (0,5 mmHg) |
MDL:
MFCD00052912 CAS-Nummer: 175205-54-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...